Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-3-octylimidazolium chloride 98%

Supplier: AMBEED, INC
Warning

A430797-100G A430797-25G
77155-916EA 151 USD InStock
77155-916 77155-918
1-Methyl-3-octylimidazolium chloride 98%
1-Methyl-3-octylimidazolium chloride
Precaución: for Research Use Only


Formula: C₁₂H₂₃ClN₂
MDL Number: MFCD03095432
CAS Number: 64697-40-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty