Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

OAC1 98%

Supplier: AMBEED, INC
Warning

A511757-100MG A511757-10MG A511757-1MG A511757-50MG A511757-5MG
77156-764EA 313 USD InStock
77156-764 77156-766 77156-768 77156-770 77156-772
OAC1 98%
OAC1
Precaución: for Research Use Only


Formula: C₁₄H₁₁N₃O
MDL Number: MFCD00572806
CAS Number: 300586-90-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty