Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Octyl-3-methylimidazolium hexafluorophosphate 98%

Supplier: AMBEED, INC
Warning

A592138-100G A592138-25G A592138-5G A592138-1G
77157-538EA 85 USD InStock
77157-538 77157-540 77157-542 77686-503
1-Octyl-3-methylimidazolium hexafluorophosphate 98%
1-Octyl-3-methylimidazolium hexafluorophosphate
Precaución: for Research Use Only


Formula: C₁₂H₂₃F₆N₂P
MDL Number: MFCD03427617
CAS Number: 304680-36-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty