Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-methylbenzaldehyde 95%

Supplier: AMBEED, INC
Warning

A819569-10G A819569-1G A819569-250MG A819569-5G
77159-342EA 307 USD InStock
77159-342 77159-344 77159-346 77159-348
3-Chloro-2-methylbenzaldehyde 95%
3-Chloro-2-methylbenzaldehyde
Precaución: for Research Use Only


Formula: C₈H₇ClO
MDL Number: MFCD11047716
CAS Number: 874-27-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty