Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-(Chloromethyl)phenyl)(phenyl)sulfane 95%

Supplier: AMBEED, INC
Danger

A106156-100G A106156-10G A106156-1G A106156-25G A106156-5G
77188-794EA 91 USD InStock
77188-794 77188-796 77188-798 77188-800 77188-802
(4-(Chloromethyl)phenyl)(phenyl)sulfane 95%
(4-(Chloromethyl)phenyl)(phenyl)sulfane
Precaución: for Research Use Only


Formula: C₁₃H₁₁ClS
MDL Number: MFCD09907764
CAS Number: 1208-87-3
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty