Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl pyrazolo[1,5-a]pyridine-3,5-dicarboxylate 97%

Supplier: AMBEED, INC
Warning

A111605-100MG A111605-250MG
77160-768EA 39 USD InStock
77160-768 77160-770
Dimethyl pyrazolo[1,5-a]pyridine-3,5-dicarboxylate 97%
Dimethyl pyrazolo[1,5-a]pyridine-3,5-dicarboxylate
Precaución: for Research Use Only


Formula: C₁₁H₁₀N₂O₄
MDL Number: MFCD20528905
CAS Number: 99446-50-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty