Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Pyridin-4-yl)-3-(quinolin-2-yl)prop-2-en-1-one 98%

Supplier: AMBEED, INC
Warning

A131123-100MG A131123-10MG A131123-1MG A131123-50MG A131123-5MG
77161-866EA 153 USD InStock
77161-866 77161-868 77161-870 77161-872 77161-874
1-(Pyridin-4-yl)-3-(quinolin-2-yl)prop-2-en-1-one 98%
1-(Pyridin-4-yl)-3-(quinolin-2-yl)prop-2-en-1-one
Precaución: for Research Use Only


Formula: C₁₇H₁₂N₂O
MDL Number: MFCD28099813
CAS Number: 4382-63-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty