Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Chloro-5-iodophenyl)(4-fluorophenyl)methanone 97%

Supplier: AMBEED, INC
Warning

A150686-100G A150686-10G A150686-1G A150686-250MG A150686-25G A150686-5G
77162-764EA 274.99 USD InStock
77162-764 77162-766 77162-768 77162-770 77162-772 77162-774
(2-Chloro-5-iodophenyl)(4-fluorophenyl)methanone 97%
(2-Chloro-5-iodophenyl)(4-fluorophenyl)methanone
Precaución: for Research Use Only


Formula: C₁₃H₇ClFIO
MDL Number: MFCD22665920
CAS Number: 915095-86-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty