Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-aminoisonicotinate 98%

Supplier: AMBEED, INC
Warning

A200968-100G A200968-10G A200968-1G A200968-25G A200968-5G
77164-592EA 227 USD InStock
77164-592 77164-594 77164-596 77164-598 77164-600
Methyl-3-aminoisonicotinate 98%
Methyl 3-aminoisonicotinate
Precaución: for Research Use Only


Formula: C₇H₈N₂O₂
MDL Number: MFCD02082535
CAS Number: 55279-30-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty