Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Octahydro-1H-4,7-methanoinden-5-ol 97%

Supplier: AMBEED, INC
Warning

A229940-1G A229940-250MG A229940-25G A229940-5G
77165-486EA 76 USD InStock
77165-486 77165-488 77165-490 77165-492
Octahydro-1H-4,7-methanoinden-5-ol 97%
Octahydro-1H-4,7-methanoinden-5-ol
Precaución: for Research Use Only


Formula: C₁₀H₁₆O
MDL Number: MFCD00168035
CAS Number: 13380-89-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty