Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Cyclopentyl-1H-pyrazol-5-amine 98%

Supplier: AMBEED, INC
Warning

A232743-1G A232743-250MG A232743-5G
77165-568EA 215 USD InStock
77165-568 77165-570 77165-572
3-Cyclopentyl-1H-pyrazol-5-amine 98%
3-Cyclopentyl-1H-pyrazol-5-amine
Precaución: for Research Use Only


Formula: C₈H₁₃N₃
MDL Number: MFCD04115105
CAS Number: 264209-16-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty