Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

HS-173 97%

Supplier: AMBEED, INC
Warning

A336342-10MG A336342-1MG A336342-25MG A336342-50MG A336342-5MG
77195-076EA 113 USD InStock
77195-076 77195-078 77195-080 77195-082 77195-084
HS-173 97%
HS-173
Precaución: for Research Use Only


Formula: C₂₁H₁₈N₄O₄S
MDL Number: MFCD28023584
CAS Number: 1276110-06-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty