Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(6-Chloropyridazin-3-yl)pivalamide 95%

Supplier: AMBEED, INC
Warning

A402716-10G A402716-1G A402716-25G A402716-5G A402716-100G
77195-944EA 109 USD InStock
77195-944 77195-946 77195-948 77195-950 77746-806
N-(6-Chloropyridazin-3-yl)pivalamide 95%
N-(6-Chloropyridazin-3-yl)pivalamide
Precaución: for Research Use Only


Formula: C₉H₁₂ClN₃O
MDL Number: MFCD17015221
CAS Number: 147362-88-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty