Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fasentin 98+%

Supplier: AMBEED, INC
Warning

A444978-10MG A444978-1MG A444978-25MG A444978-50MG A444978-5MG
77196-416EA 149 USD InStock
77196-416 77196-418 77196-420 77196-422 77196-424
Fasentin 98+%
Fasentin
Precaución: for Research Use Only


Formula: C₁₁H₉ClF₃NO₂
MW: 279.64 g/mol
MDL Number: MFCD01001285
CAS Number: 392721-37-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty