Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethoxytriphenylsilane 95%

Supplier: AMBEED, INC
Warning

A459951-100G A459951-1G A459951-25G A459951-5G
77168-946EA 374 USD InStock
77168-946 77168-948 77168-950 77168-952
Ethoxytriphenylsilane 95%
Ethoxytriphenylsilane
Precaución: for Research Use Only


Formula: C₂₀H₂₀OSi
MDL Number: MFCD00053752
CAS Number: 1516-80-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty