Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[(6-Chloro[1,2,4]triazolo[4,3-b]pyridazin-3-yl)methyl]amine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A499690-100MG A499690-250MG
77197-010EA 87 USD InStock
77197-010 77197-012
[(6-Chloro[1,2,4]triazolo[4,3-b]pyridazin-3-yl)methyl]amine hydrochloride 97%
[(6-Chloro[1,2,4]triazolo[4,3-b]pyridazin-3-yl)methyl]amine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₇Cl₂N₅
MDL Number: MFCD19103499
CAS Number: 518050-85-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty