Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Allyl-2-methylbenzene 95%

Supplier: AMBEED, INC
Danger

A506839-1G A506839-25G A506839-5G A506839-250MG
77197-078EA 29 USD InStock
77197-078 77197-080 77197-082 77747-045
1-Allyl-2-methylbenzene 95%
1-Allyl-2-methylbenzene
Precaución: for Research Use Only


Formula: C₁₀H₁₂
MDL Number: MFCD00060911
CAS Number: 1587-04-8
UN: 2924
ADR: 3,8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty