Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 6-ethynylnicotinate 97%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A562410-250MG A562410-100MG A562410-1G
77169-828EA 24 USD InStock
77169-828 77684-412 77684-413
Methyl 6-ethynylnicotinate 97%, Ambeed.Inc
Methyl 6-ethynylnicotinate
Precaución: For research use only.


Formula: C₉H₇NO₂
MW: 161.16 g/mol
MDL Number: MFCD16988751
CAS Number: 216444-00-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty