Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Acetylpiperidine-4-carboxylic acid 95%

Supplier: AMBEED, INC
Warning

A593200-100G A593200-10G A593200-1G A593200-25G A593200-5G
77197-912EA 61 USD InStock
77197-912 77197-914 77197-916 77197-918 77197-920
1-Acetylpiperidine-4-carboxylic acid 95%
1-Acetylpiperidine-4-carboxylic acid
Precaución: for Research Use Only


Formula: C₈H₁₃NO₃
MDL Number: MFCD00023706
CAS Number: 25503-90-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty