Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-2-Methoxy-4-(prop-1-en-1-yl)phenyl acetate 98+%

Supplier: AMBEED, INC
Warning

A599434-100G A599434-25G
77197-998EA 51 USD InStock
77197-998 77198-000 77198-002
(E)-2-Methoxy-4-(prop-1-en-1-yl)phenyl acetate 98+%
(E)-2-Methoxy-4-(prop-1-en-1-yl)phenyl acetate
Precaución: for Research Use Only


Formula: C₁₂H₁₄O₃
MDL Number: MFCD00026984
CAS Number: 5912-87-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty