Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-(tert-butyl)-1-iodobenzene 95%

Supplier: AMBEED, INC
Warning

A606825-10G A606825-1G A606825-250MG A606825-5G
77198-058EA 63 USD InStock
77198-058 77198-060 77198-062 77198-064
2-Bromo-4-(tert-butyl)-1-iodobenzene 95%
2-Bromo-4-(tert-butyl)-1-iodobenzene
Precaución: for Research Use Only


Formula: C₁₀H₁₂BrI
MDL Number: MFCD09878146
CAS Number: 860435-39-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty