Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-3-(trifluoromethyl)phenol 97%

Supplier: AMBEED, INC
Danger

A856704-1G A856704-250MG A856704-25G A856704-5G
77171-664EA 36 USD InStock
77171-664 77171-666 77171-668 77171-670
2-Methyl-3-(trifluoromethyl)phenol 97%
2-Methyl-3-(trifluoromethyl)phenol
Precaución: for Research Use Only


Formula: C₈H₇F₃O
MDL Number: MFCD09832280
CAS Number: 934180-47-9
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty