1-Boc-4-(5-bromopyrimidin-2-yl)piperazine 98%

Supplier: AMBEED, INC
Warning

A877764-100G A877764-10G A877764-1G A877764-250MG A877764-25G A877764-5G
77171-742EA 110 USD
77171-742 77171-744 77171-746 77171-748 77171-750 77171-752
1-Boc-4-(5-bromopyrimidin-2-yl)piperazine 98%
1-Boc-4-(5-bromopyrimidin-2-yl)piperazine

Precaución: for Research Use Only

Formula: C₁₃H₁₉BrN₄O₂
MW: 343.23 g/mol
MDL Number: MFCD07375175
CAS Number: 374930-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR