Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-4-methylphenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A878589-100G A878589-10G A878589-1G A878589-25G A878589-5G
77200-130EA 302 USD InStock
77200-130 77200-132 77200-134 77200-136 77200-138
3-Chloro-4-methylphenylboronic acid 97%
3-Chloro-4-methylphenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₈BClO₂
MDL Number: MFCD04039010
CAS Number: 175883-63-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty