Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2,3-Trifluorobenzene 98%

Supplier: AMBEED, INC
Danger

A905443-10G A905443-25G A905443-5G
77200-234EA 11 USD InStock
77200-234 77200-236 77200-238
1,2,3-Trifluorobenzene 98%
1,2,3-Trifluorobenzene
Precaución: for Research Use Only


Formula: C₆H₃F₃
MDL Number: MFCD00012013
CAS Number: 1489-53-8
UN: 1993
ADR: 3,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty