Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl azetidine-3-carboxylate hydrochloride 97%

Supplier: AMBEED, INC
Warning

A112952-100MG A112952-250MG
77233-672EA 32 USD InStock
77233-672 77233-674
tert-Butyl azetidine-3-carboxylate hydrochloride 97%
tert-Butyl azetidine-3-carboxylate hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₅NO₂·ClH
MDL Number: MFCD20528798
CAS Number: 53871-08-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty