5-(Trifluoromethyl)indoline 97%

Supplier: AMBEED, INC
Warning

A113909-100MG A113909-1G A113909-250MG A113909-5G
77271-100EA 32 USD
77271-100 77271-102 77271-104 77271-106
5-(Trifluoromethyl)indoline 97%
5-(Trifluoromethyl)indoline

Precaución: for Research Use Only

Formula: C₉H₈F₃N
MW: 187.162 g/mol
MDL Number: MFCD03095322
CAS Number: 162100-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR