2,2'-Oxybis(ethylamine) dihydrochloride 98%
Supplier: AMBEED, INC
|
Warning
|
A122235-1G
A122235-5G
A122235-250MG
77246-158EA
27
USD
77246-158
77246-160
77745-896
2,2'-Oxybis(ethylamine) dihydrochloride 98%
2,2'-Oxybis(ethylamine) dihydrochloride
Precaución: for Research Use Only
Formula:
C₄H₁₂N₂O MW: 177.07 g/mol |
MDL Number:
MFCD00012891 CAS Number: 60792-79-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...