Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methylnicotinic acid hydrochloride 95%

Supplier: AMBEED, INC
Warning

A123053-10G A123053-250MG A123053-5G A123053-1G
77284-944EA 210.01 USD InStock
77284-944 77284-946 77284-948 77747-890
4-Methylnicotinic acid hydrochloride 95%
4-Methylnicotinic acid hydrochloride
Precaución: for Research Use Only


Formula: C₇H₈ClNO₂
MDL Number: MFCD00832700
CAS Number: 94015-05-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty