Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-formylphenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A125673-10G A125673-1G A125673-250MG A125673-25G A125673-5G
77271-766EA 12 USD InStock
77271-766 77271-768 77271-770 77271-772 77271-774
2-Chloro-5-formylphenylboronic acid 97%
2-Chloro-5-formylphenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₆BClO₃
MDL Number: MFCD06656269
CAS Number: 1150114-78-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty