Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-4-hydroxyphenylacetic acid 98%

Supplier: AMBEED, INC
Warning

A126613-10G A126613-1G A126613-25G A126613-5G
77234-336EA 68 USD InStock
77234-336 77234-338 77234-340 77234-342
3-Bromo-4-hydroxyphenylacetic acid 98%
3-Bromo-4-hydroxyphenylacetic acid
Precaución: for Research Use Only


Formula: C₈H₇BrO₃
MDL Number: MFCD00016833
CAS Number: 38692-80-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty