Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-methyl-4-nitrobenzoate 98%

Supplier: AMBEED, INC
Warning

A127769-100G A127769-10G A127769-25G A127769-500G A127769-5G
77271-850EA 30 USD InStock
77271-850 77271-852 77271-854 77271-856 77271-858
Methyl-3-methyl-4-nitrobenzoate 98%
Methyl 3-methyl-4-nitrobenzoate
Precaución: for Research Use Only


Formula: C₉H₉NO₄
MW: 195.18 g/mol
MDL Number: MFCD00085640
CAS Number: 24078-21-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty