Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-ethoxypyridine 97%

Supplier: AMBEED, INC
Warning

A127835-100MG A127835-1G A127835-250MG A127835-5G
77258-322EA 6 USD InStock
77258-322 77258-324 77258-326 77258-328
2-Chloro-4-ethoxypyridine 97%
2-Chloro-4-ethoxypyridine
Precaución: for Research Use Only


Formula: C₇H₈ClNO
MW: 157.598 g/mol
MDL Number: MFCD00234316
CAS Number: 52311-50-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty