Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzo[b]furan-5-boronic acid 97%

Supplier: AMBEED, INC
Warning

A129428-100MG A129428-1G A129428-250MG A129428-5G
77234-438EA 10 USD InStock
77234-438 77234-440 77234-442 77745-934
Benzo[b]furan-5-boronic acid 97%
Benzo[b]furan-5-boronic acid
Precaución: for Research Use Only


Formula: C₈H₇BO₃
MDL Number: MFCD04038999
CAS Number: 331834-13-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty