Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[1,1'-Biphenyl]-3,3',5,5'-tetracarbaldehyde 98%

Supplier: AMBEED, INC
Warning

A133250-100MG A133250-1G A133250-250MG
77258-546EA 29 USD InStock
77258-546 77258-548 77258-550
[1,1'-Biphenyl]-3,3',5,5'-tetracarbaldehyde 98%
[1,1'-Biphenyl]-3,3',5,5'-tetracarbaldehyde
Precaución: for Research Use Only


Formula: C₁₆H₁₀O₄
MDL Number: MFCD31700813
CAS Number: 150443-85-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty