Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluorophenetole 97%

Supplier: AMBEED, INC
Warning

A138096-100G A138096-10G A138096-1G A138096-25G A138096-5G
77272-346EA 182 USD InStock
77272-346 77272-348 77272-350 77272-352 77272-354
3-Fluorophenetole 97%
3-Fluorophenetole
Precaución: for Research Use Only


Formula: C₈H₉FO
MDL Number: MFCD00673011
CAS Number: 458-03-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty