Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-3-nitropyridine 95%

Supplier: AMBEED, INC
Warning

A140899-100G A140899-10G A140899-25G A140899-500G A140899-5G
77285-732EA 53 USD InStock
77285-732 77285-734 77285-736 77285-738 77285-740
4-Hydroxy-3-nitropyridine 95%
4-Hydroxy-3-nitropyridine
Precaución: for Research Use Only


Formula: C₅H₄N₂O₃
MDL Number: MFCD00128874
CAS Number: 5435-54-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty