Ethyl-1-methyl-3-piperidinecarboxylate 97%

Supplier: AMBEED, INC
Warning

A149916-100G A149916-10G A149916-25G
77259-208EA 72 USD
77259-208 77259-210 77259-212
Ethyl-1-methyl-3-piperidinecarboxylate 97%
Ethyl-1-methyl-3-piperidinecarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₇NO₂
MW: 171.24 g/mol
MDL Number: MFCD00006496
CAS Number: 5166-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR