Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Bromopyridin-4-yl)ethanone 95%

Supplier: AMBEED, INC
Warning

A152590-100MG A152590-10G A152590-1G A152590-250MG A152590-5G
77259-310EA 15 USD InStock
77259-310 77259-312 77259-314 77259-316 77259-318
1-(2-Bromopyridin-4-yl)ethanone 95%
1-(2-Bromopyridin-4-yl)ethanone
Precaución: for Research Use Only


Formula: C₇H₆BrNO
MDL Number: MFCD07367876
CAS Number: 864674-02-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty