Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Geranyllinalool (mixture of isomers) 97%

Supplier: AMBEED, INC
Warning

A154408-100G A154408-10G A154408-1G A154408-25G A154408-5G
77286-216EA 191 USD InStock
77286-216 77286-218 77286-220 77286-222 77286-224
Geranyllinalool (mixture of isomers) 97%
Geranyllinalool (mixture of isomers)
Precaución: for Research Use Only


Formula: C₂₀H₃₄O
MDL Number: MFCD00059363
CAS Number: 68931-30-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty