Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3'-Dithiodipropionic acid 98%

Supplier: AMBEED, INC
Warning

A161411-100G A161411-25G
77273-326EA 31 USD InStock
77273-326 77273-328 77273-330
3,3'-Dithiodipropionic acid 98%
3,3'-Dithiodipropionic acid
Precaución: for Research Use Only


Formula: C₆H₁₀O₄S₂
MDL Number: MFCD00002780
CAS Number: 1119-62-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty