Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3,5-dimethyl-1H-pyrrole-2-carbaldehyde 95%

Supplier: AMBEED, INC
Warning

A163271-100MG A163271-1G A163271-250MG A163271-5G
77286-528EA 39 USD InStock
77286-528 77286-530 77286-532 77286-534
4-Bromo-3,5-dimethyl-1H-pyrrole-2-carbaldehyde 95%
4-Bromo-3,5-dimethyl-1H-pyrrole-2-carbaldehyde
Precaución: for Research Use Only


Formula: C₇H₈BrNO
MDL Number: MFCD08234561
CAS Number: 89909-51-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty