Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(-)-2-Hydroxy-1,2,2-triphenylethyl acetate 95%

Supplier: AMBEED, INC
Warning

A168601-1G A168601-250MG A168601-5G
77286-720EA 64 USD InStock
77286-720 77286-722 77286-724
(S)-(-)-2-Hydroxy-1,2,2-triphenylethyl acetate 95%
(S)-(-)-2-Hydroxy-1,2,2-triphenylethyl acetate
Precaución: for Research Use Only


Formula: C₂₂H₂₀O₃
MDL Number: MFCD00066241
CAS Number: 95061-51-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty