Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,7-Bis(4-methoxybenzyl)-3,7-dihydro-2H-diimidazo[4,5-d:4',5'-f][1,3]diazepin-2-one 98%

Supplier: AMBEED, INC
Warning

A179326-10MG A179326-1MG A179326-25MG A179326-2MG A179326-5MG A179326-50MG
77260-158EA 67 USD InStock
77260-158 77260-160 77260-162 77260-164 77260-166 77748-171
3,7-Bis(4-methoxybenzyl)-3,7-dihydro-2H-diimidazo[4,5-d:4',5'-f][1,3]diazepin-2-one 98%
3,7-Bis(4-methoxybenzyl)-3,7-dihydro-2H-diimidazo[4,5-d:4',5'-f][1,3]diazepin-2-one
Precaución: for Research Use Only


Formula: C₂₃H₂₀N₆O₃
MDL Number: MFCD30489740
CAS Number: 1070773-09-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty