Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Chloropropyl)benzene 95%

Supplier: AMBEED, INC
Warning

A184642-100G A184642-25G A184642-500G A184642-5G A184642-1000G
77236-664EA 44 USD InStock
77236-664 77236-666 77236-668 77236-670 77746-192
(3-Chloropropyl)benzene 95%
(3-Chloropropyl)benzene
Precaución: for Research Use Only


Formula: ClH₂CCH₂CH₂(C₆H₅)
MDL Number: MFCD00001001
CAS Number: 104-52-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty