Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Isopropyl-4-methylpyridin-3-amine 97%

Supplier: AMBEED, INC
Warning

A202292-100MG A202292-1G A202292-250MG A202292-25G A202292-5G
77260-804EA 5 USD InStock
77260-804 77260-806 77260-808 77260-810 77260-812
2-Isopropyl-4-methylpyridin-3-amine 97%
2-Isopropyl-4-methylpyridin-3-amine
Precaución: for Research Use Only


Formula: C₉H₁₄N₂
MDL Number: MFCD30828926
CAS Number: 1698293-93-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty