Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dimethylbenzoic acid 95%

Supplier: AMBEED, INC
Warning

A204196-1000G A204196-10G A204196-25G A204196-500G A204196-100G
77274-938EA 61 USD InStock
77274-938 77274-940 77274-942 77274-944 77748-293
3,5-Dimethylbenzoic acid 95%
3,5-Dimethylbenzoic acid
Precaución: for Research Use Only


Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 278
MDL Number: MFCD00002525
CAS Number: 499-06-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty