Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Difluoro-6-nitroaniline 98%

Supplier: AMBEED, INC
Warning

A217622-100G A217622-10G A217622-1G A217622-25G A217622-5G
77249-272EA 145 USD InStock
77249-272 77249-274 77249-276 77249-278 77249-280
2,4-Difluoro-6-nitroaniline 98%
2,4-Difluoro-6-nitroaniline
Precaución: for Research Use Only


Formula: C₆H₄F₂N₂O₂
MDL Number: MFCD00034058
CAS Number: 364-30-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty