Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dichloro-4-fluoronitrobenzene 98%

Supplier: AMBEED, INC
Warning

A238333-100MG A238333-10G A238333-1G A238333-250MG A238333-5G
77249-718EA 7 USD InStock
77249-718 77249-720 77249-722 77249-724 77249-726
2,3-Dichloro-4-fluoronitrobenzene 98%
2,3-Dichloro-4-fluoronitrobenzene
Precaución: for Research Use Only


Formula: C₆H₂Cl₂FNO₂
MDL Number: MFCD09763598
CAS Number: 36556-51-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty