3,5-Dimethoxybenzonitrile 97%

Supplier: AMBEED, INC
Warning

A239716-100G A239716-10G A239716-5G A239716-1G
77275-986EA 111 USD
77275-986 77275-988 77275-990 77748-417
3,5-Dimethoxybenzonitrile 97%
3,5-Dimethoxybenzonitrile

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MW: 163.18
Boiling Pt: 283
Melting Pt: 92
MDL Number: MFCD00001804
CAS Number: 19179-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR